siRNA Details
| Gene Symbol: Cep89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028120.2 | NP_082396.1 | 14 | 1627 | ||
| GenBank mRNA | AK012067.1 | - | 14 | 1628 | ||
| AK029107.1 | - | 14 | 1658 | |||
| BC003981.1 | - | 14 | 1682 | 3490246 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cep89| Gene Symbol: Cep89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028120.2 | NP_082396.1 | 14 | 1627 | ||
| GenBank mRNA | AK012067.1 | - | 14 | 1628 | ||
| AK029107.1 | - | 14 | 1658 | |||
| BC003981.1 | - | 14 | 1682 | 3490246 | ||