siRNA Details
| Gene Symbol: Slc37a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028123.3 | NP_082399.3 | 10 | 1068 | ||
| GenBank mRNA | AK012071.1 | - | 2 | 291 | ||
| AK076213.1 | - | 10 | 1011 | |||
| AK077836.1 | - | 10 | 1012 | |||
| AK145242.1 | - | 10 | 1038 | |||
| AK167716.1 | - | 10 | 1024 | |||
| BC005744.1 | - | 10 | 887 | |||
| BC099673.1 | - | 11 | 1005 | 4936456 | ||
| BC100391.1 | - | 11 | 1102 | |||

Slc37a3
Genomic Map