siRNA Details
| Gene Symbol: Slc37a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC005744.1 | - | 15 | 1403 | ||
| BC099673.1 | - | 16 | 1521 | 4936456 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc37a3| Gene Symbol: Slc37a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC005744.1 | - | 15 | 1403 | ||
| BC099673.1 | - | 16 | 1521 | 4936456 | ||