siRNA Details
| Gene Symbol: Slc37a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028123.3 | NP_082399.3 | 12 | 1319 | ||
| GenBank mRNA | AK012071.1 | - | 4 | 542 | ||
| AK076213.1 | - | 12 | 1262 | |||
| AK077836.1 | - | 12 | 1262 | |||
| AK145242.1 | - | 12 | 1289 | |||
| AK167716.1 | - | 12 | 1275 | |||
| BC005744.1 | - | 12 | 1138 | |||
| BC099673.1 | - | 13 | 1256 | 4936456 | ||
| BC100391.1 | - | 13 | 1353 | |||

Slc37a3
Genomic Map