siRNA Details
| Gene Symbol: Trim29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023655.2 | NP_076144.2 | 7 | 1683 | ||
| GenBank mRNA | AK013219.1 | - | 6 | 734 | ||
| AK028448.1 | - | 7 | 916 | |||
| BC027353.1 | - | 7 | 1598 | 3988926 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim29| Gene Symbol: Trim29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023655.2 | NP_076144.2 | 7 | 1683 | ||
| GenBank mRNA | AK013219.1 | - | 6 | 734 | ||
| AK028448.1 | - | 7 | 916 | |||
| BC027353.1 | - | 7 | 1598 | 3988926 | ||