siRNA Details
Gene Symbol: Atxn7l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028139.5 | NP_082415.2 | 3, 4 | 405 | ||
GenBank mRNA | AK013145.1 | - | 3, 4 | 312 | ||
BC017546.1 | - | 2, 3 | 206 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Atxn7l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028139.5 | NP_082415.2 | 3, 4 | 405 | ||
GenBank mRNA | AK013145.1 | - | 3, 4 | 312 | ||
BC017546.1 | - | 2, 3 | 206 |