siRNA Details
| Gene Symbol: Mfsd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028140.4 | NP_082416.2 | 2 | 228 | ||
| GenBank mRNA | AK013144.1 | - | 2 | 120 | ||
| AK032080.1 | - | 2 | 249 | |||
| AK035291.1 | - | 2 | 191 | |||
| AK077957.1 | - | 2 | 229 | |||
| BC113183.1 | - | 2 | 159 | 40061891 | ||
| BC113790.1 | - | 2 | 159 | 40061889 | ||

Mfsd8
Genomic Map