siRNA Details
| Gene Symbol: Snx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026998.3 | NP_081274.2 | 6, 7 | 629 | ||
| GenBank mRNA | AK008135.1 | - | 1, 2 | 94 | ||
| AK011640.1 | - | 7, 8 | 631 | |||
| AK013158.1 | - | 6, 7 | 630 | |||
| BC025911.1 | - | 6, 7 | 616 | |||
| BC061028.1 | - | 7, 8 | 670 | 6594585 | ||

Snx6
Genomic Map