siRNA Details
| Gene Symbol: 2510009E07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001881.2 | NP_001001881.1 | 2 | 757 | ||
| GenBank mRNA | AK044207.1 | - | 2 | 675 | ||
| BC072629.1 | - | 2 | 706 | 6833156 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2510009E07Rik| Gene Symbol: 2510009E07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001881.2 | NP_001001881.1 | 2 | 757 | ||
| GenBank mRNA | AK044207.1 | - | 2 | 675 | ||
| BC072629.1 | - | 2 | 706 | 6833156 | ||