siRNA Details
Gene Symbol: Skiv2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028151.2 | NP_082427.1 | 14 | 1526 | ||
GenBank mRNA | AK012158.1 | - | 14 | 1526 | ||
AK078091.1 | - | 14 | 1545 | |||
AK088426.1 | - | 13 | 1363 | |||
AK129045.1 | - | 7 | 681 | |||
BC014810.1 | - | 6 | 560 | |||
BC029230.1 | - | 14 | 1516 | 4488921 |
