siRNA Details
| Gene Symbol: Otud6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152812.3 | NP_690025.2 | 3, 4 | 527 | ||
| GenBank mRNA | AK144761.1 | - | 3, 4 | 350 | ||
| AK155761.1 | - | 3, 4 | 378 | |||
| AK166445.1 | - | 3, 4 | 357 | |||
| AK166546.1 | - | 3, 4 | 354 | |||
| AK169134.1 | - | 3, 4 | 342 | |||
| AK172229.1 | - | 3, 4 | 332 | |||
| AK172338.1 | - | 3, 4 | 332 | |||
| BC031474.1 | - | 3, 4 | 333 | 2599116 | ||
| BC087552.1 | - | 3, 4 | 324 | 6331952 | ||
| CN724613.1 | - | 3 | 527 | |||

Otud6b
Genomic Map