siRNA Details
| Gene Symbol: 1700001P01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028156.2 | NP_082432.2 | 2 | 336 | ||
| GenBank mRNA | AK005619.1 | - | 2 | 300 | ||
| BC048607.1 | - | 2 | 336 | 6741651 | ||
| CA466212.1 | - | 304 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700001P01Rik| Gene Symbol: 1700001P01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028156.2 | NP_082432.2 | 2 | 336 | ||
| GenBank mRNA | AK005619.1 | - | 2 | 300 | ||
| BC048607.1 | - | 2 | 336 | 6741651 | ||
| CA466212.1 | - | 304 | ||||