siRNA Details
| Gene Symbol: 1700021F07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028158.1 | NP_082434.1 | 4 | 399 | ||
| GenBank mRNA | AK006200.1 | - | 4 | 358 | ||
| BC050790.1 | - | 4 | 391 | 6741974 | ||
| CN835193.1 | - | 4 | 399 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700021F07Rik| Gene Symbol: 1700021F07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028158.1 | NP_082434.1 | 4 | 399 | ||
| GenBank mRNA | AK006200.1 | - | 4 | 358 | ||
| BC050790.1 | - | 4 | 391 | 6741974 | ||
| CN835193.1 | - | 4 | 399 | |||