siRNA Details
| Gene Symbol: Psg21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027403.4 | NP_081679.2 | 2 | 425 | ||
| GenBank mRNA | AK005492.1 | - | 2 | 423 | ||
| AK005545.1 | - | 2 | 423 | |||
| AK131670.1 | - | 2 | 423 | |||
| BC100324.1 | - | 2 | 402 | 30715195 | ||
| BC100325.1 | - | 2 | 402 | 30787148 | ||

Psg21
Genomic Map