siRNA Details
| Gene Symbol: 1700030K09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028170.2 | NP_082446.2 | 4 | 1172 | ||
| GenBank mRNA | AK089918.1 | - | 4 | 1114 | ||
| AK133815.1 | - | 4 | 1176 | |||
| AK157359.1 | - | 3 | 383 | |||
| AK172202.1 | - | 3 | 381 | |||
| BC008558.1 | - | 4 | 1126 | 3592293 | ||

1700030K09Rik
Genomic Map