siRNA Details
| Gene Symbol: Sh2d4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028182.1 | NP_082458.1 | 5, 6 | 1164 | ||
| GenBank mRNA | AK008803.1 | - | 5, 6 | 950 | ||
| AK157396.1 | - | 6, 7 | 962 | |||
| AX776203.1 | - | 4, 5 | 621 | |||
| BC023784.1 | - | 5, 6 | 557 | |||
| BC116682.1 | - | 5, 6 | 872 | 40106612 | ||

Sh2d4a
Genomic Map