siRNA Details
| Gene Symbol: Nkd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028186.4 | NP_082462.3 | 4 | 416 | ||
| GenBank mRNA | AF358136.1 | - | 4 | 366 | ||
| AK008814.1 | - | 4 | 398 | |||
| AK081904.1 | - | 3 | 362 | |||
| AK145155.1 | - | 4 | 398 | |||
| AK146667.1 | - | 4 | 398 | |||
| AK158350.1 | - | 4 | 398 | |||
| AK165116.1 | - | 3 | 362 | |||
| AK199881.1 | - | 2 | 78 | |||
| AK203955.1 | - | 2 | 79 | |||
| BC019952.1 | - | 4 | 385 | 4021772 | ||

Nkd2
Genomic Map