siRNA Details
| Gene Symbol: Nkd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028186.4 | NP_082462.3 | 7 | 685 | ||
| GenBank mRNA | AF358136.1 | - | 7 | 635 | ||
| AK008814.1 | - | 7 | 667 | |||
| AK081904.1 | - | 6 | 631 | |||
| AK145155.1 | - | 7 | 667 | |||
| AK146667.1 | - | 7 | 667 | |||
| AK158350.1 | - | 7 | 667 | |||
| AK165116.1 | - | 6 | 631 | |||
| BC019952.1 | - | 7 | 654 | 4021772 | ||

Nkd2
Genomic Map