siRNA Details
| Gene Symbol: Rusc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083807.1 | NP_001077276.1 | 6, 7 | 1988 | ||
| NM_001083808.1 | NP_001077277.1 | 5, 6 | 528 | |||
| NM_028188.2 | NP_082464.2 | 7, 8 | 2399 | |||
| GenBank mRNA | AK032334.1 | - | 5, 6 | 1018 | ||
| AK039664.1 | - | 7, 8 | 2327 | |||
| AK042689.1 | - | 6, 7 | 1936 | |||
| AK170012.1 | - | 5, 6 | 542 | |||
| AK170210.1 | - | 5, 6 | 491 | |||
| BC052081.1 | - | 4, 5 | 722 | |||
| BC056360.1 | - | 6, 7 | 1926 | 6816267 | ||
| BC057034.1 | - | 5, 6 | 594 | 5687517 | ||

Rusc1
Genomic Map