siRNA Details
Gene Symbol: Zfp777 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081382.1 | NP_001074851.1 | 2 | 325 | ||
GenBank mRNA | BC066046.1 | - | 2 | 415 | ||
BC150871.1 | - | 2 | 349 | 9087782 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp777 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081382.1 | NP_001074851.1 | 2 | 325 | ||
GenBank mRNA | BC066046.1 | - | 2 | 415 | ||
BC150871.1 | - | 2 | 349 | 9087782 |