siRNA Details
| Gene Symbol: Zfp777 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081382.1 | NP_001074851.1 | 2 | 826 | ||
| GenBank mRNA | BC066046.1 | - | 2 | 916 | ||
| BC150871.1 | - | 2 | 850 | 9087782 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp777| Gene Symbol: Zfp777 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081382.1 | NP_001074851.1 | 2 | 826 | ||
| GenBank mRNA | BC066046.1 | - | 2 | 916 | ||
| BC150871.1 | - | 2 | 850 | 9087782 | ||