siRNA Details
| Gene Symbol: 2510002D24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033164.2 | NP_001028336.1 | 3 | 382 | ||
| GenBank mRNA | AK153794.1 | - | 3 | 373 | ||
| AK160192.1 | - | 3 | 385 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2510002D24Rik| Gene Symbol: 2510002D24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033164.2 | NP_001028336.1 | 3 | 382 | ||
| GenBank mRNA | AK153794.1 | - | 3 | 373 | ||
| AK160192.1 | - | 3 | 385 | |||