siRNA Details
| Gene Symbol: Brf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028193.3 | NP_082469.2 | 8 | 1088 | ||
| GenBank mRNA | BC037224.1 | - | 8 | 871 | 3257364 | |
| CF737042.1 | - | 5 | 409 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Brf1| Gene Symbol: Brf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028193.3 | NP_082469.2 | 8 | 1088 | ||
| GenBank mRNA | BC037224.1 | - | 8 | 871 | 3257364 | |
| CF737042.1 | - | 5 | 409 | |||