siRNA Details
| Gene Symbol: Tmem158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002267.2 | NP_001002267.2 | 1 | 1609 | ||
| GenBank mRNA | AK009669.1 | - | 1 | 1584 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem158| Gene Symbol: Tmem158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002267.2 | NP_001002267.2 | 1 | 1609 | ||
| GenBank mRNA | AK009669.1 | - | 1 | 1584 | ||