siRNA Details
| Gene Symbol: Nkg7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024253.4 | NP_077215.2 | 2 | 443 | ||
| GenBank mRNA | AK010872.1 | - | 2 | 443 | ||
| AK088383.1 | - | 2 | 452 | |||
| BC055882.1 | - | 2 | 508 | 3497559 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nkg7| Gene Symbol: Nkg7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024253.4 | NP_077215.2 | 2 | 443 | ||
| GenBank mRNA | AK010872.1 | - | 2 | 443 | ||
| AK088383.1 | - | 2 | 452 | |||
| BC055882.1 | - | 2 | 508 | 3497559 | ||