siRNA Details
| Gene Symbol: Kif1bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028197.2 | NP_082473.2 | 7 | 1849 | ||
| GenBank mRNA | AK010898.1 | - | 1 | 102 | ||
| AK129320.1 | - | 7 | 1834 | |||
| AK141324.1 | - | 7 | 1858 | |||
| AK149629.1 | - | 7 | 1787 | |||
| AK158730.1 | - | 8 | 1583 | |||
| AK185659.1 | - | 1 | 138 | |||
| AK215072.1 | - | 1 | 137 | |||
| BC038828.1 | - | 3 | 1158 | |||
| BC046485.1 | - | 7 | 1820 | 5350638 | ||
| BC055307.1 | - | 7 | 1803 | 6493547 | ||

Kif1bp
Genomic Map