siRNA Details
| Gene Symbol: Xpo5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028198.2 | NP_082474.1 | 10 | 1096 | ||
| GenBank mRNA | AF343581.1 | - | 11 | 1096 | ||
| AK010389.1 | - | 10 | 1127 | |||
| BC039187.1 | - | 23 | ||||
| BC092371.1 | - | 1169 | ||||
| BC131660.1 | - | 10 | 1092 | 40110670 | ||
| BC131661.1 | - | 10 | 1092 | 40110672 | ||

Xpo5
Genomic Map