siRNA Details
| Gene Symbol: Asb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133346.2 | NP_579924.1 | 4, 5 | 623 | ||
| GenBank mRNA | AF398967.1 | - | 2661 | |||
| AK010909.1 | - | 4, 5 | 604 | |||
| AK049312.1 | - | 4, 5 | 612 | |||
| AK088890.1 | - | 4, 5 | 558 | |||
| AK190893.1 | - | 1, 2 | 101 | |||
| AY665653.1 | - | 4, 5 | 509 | |||
| BC019192.1 | - | 4, 5 | 615 | 5035395 | ||

Asb6
Genomic Map