siRNA Details
| Gene Symbol: Asb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133346.2 | NP_579924.1 | 6 | 1012 | ||
| GenBank mRNA | AF398967.1 | - | 3050 | |||
| AK010909.1 | - | 6 | 993 | |||
| AK049312.1 | - | 6 | 1001 | |||
| AK088890.1 | - | 6 | 947 | |||
| AY665653.1 | - | 6 | 898 | |||
| BC019192.1 | - | 6 | 1004 | 5035395 | ||
| BC085485.1 | - | 5 | 1693 | |||

Asb6
Genomic Map