siRNA Details
| Gene Symbol: Plxdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163608.1 | NP_001157080.1 | 5, 6 | 755 | ||
| NM_028199.3 | NP_082475.3 | 5, 6 | 734 | |||
| GenBank mRNA | AF378760.1 | - | 5, 6 | 668 | ||
| AK010361.1 | - | 5, 6 | 734 | |||
| AK036144.1 | - | 5, 6 | 709 | |||
| BC114352.2 | - | 5, 6 | 654 | 40057410 | ||

Plxdc1
Genomic Map