siRNA Details
| Gene Symbol: Vps9d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028200.1 | NP_082476.1 | 12 | 1578 | ||
| GenBank mRNA | AK005046.2 | - | 12 | 1530 | ||
| AK010402.1 | - | 12 | 1511 | |||
| AK028751.1 | - | 12 | 1521 | |||
| AK032561.1 | - | 11 | 2035 | |||
| AK151498.1 | - | 12 | 1584 | |||
| AK154409.1 | - | 12 | 1496 | |||
| AK155433.1 | - | 12 | 1496 | |||
| AK210076.1 | - | 1 | 17 | |||
| AY751521.1 | - | 12 | 1751 | |||
| BC057621.1 | - | 12 | 1579 | 6402384 | ||

Vps9d1
Genomic Map