siRNA Details
| Gene Symbol: Klhl40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028202.3 | NP_082478.1 | 2 | 1437 | ||
| GenBank mRNA | AK078669.1 | - | 2 | 1351 | ||
| AK160799.1 | - | 2 | 1297 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhl40| Gene Symbol: Klhl40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028202.3 | NP_082478.1 | 2 | 1437 | ||
| GenBank mRNA | AK078669.1 | - | 2 | 1351 | ||
| AK160799.1 | - | 2 | 1297 | |||