siRNA Details
| Gene Symbol: Zc2hc1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172414.4 | NP_766002.2 | 3 | 1578 | ||
| GenBank mRNA | AK032455.1 | - | 3 | 1482 | ||
| AK033213.1 | - | 3 | 1473 | |||
| AK036602.1 | - | 3 | 1516 | |||
| BC049940.1 | - | 3 | 1492 | 6305959 | ||

Zc2hc1c
Genomic Map