siRNA Details
| Gene Symbol: Cdpf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164625.1 | NP_001158097.1 | 4 | 398 | ||
| NM_001164626.1 | NP_001158098.1 | 3 | 192 | |||
| NM_197998.2 | NP_932115.2 | 4 | 339 | |||
| GenBank mRNA | AK008763.1 | - | 1 | 26 | ||
| BC025858.1 | - | 4 | 301 | 5007877 | ||
| BQ550379.1 | - | 4 | 357 | |||
| BY081241.1 | - | 4 | 301 | |||

Cdpf1
Genomic Map