siRNA Details
Gene Symbol: 2210016L21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028211.1 | NP_082487.1 | 2 | 168 | ||
GenBank mRNA | AK008740.1 | - | 2 | 166 | ||
AK135000.1 | - | 186 | ||||
AK159360.1 | - | 2 | 166 | |||
BC031162.1 | - | 2 | 168 | 4982352 |
