siRNA Details
| Gene Symbol: 2210408I21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081353.1 | NP_001074822.1 | 4 | 609 | ||
| NM_001145676.1 | NP_001139148.1 | 4 | 609 | |||
| GenBank mRNA | AK050886.1 | - | 4 | 595 | ||
| AK143619.1 | - | 4 | 547 | |||
| AK165312.1 | - | 4 | 606 | |||
| BC089469.1 | - | 4 | 591 | |||
| BC151066.1 | - | 4 | 591 | 9087979 | ||
| BC157949.1 | - | 4 | 591 | 9088124 | ||

2210408I21Rik
Genomic Map