siRNA Details
| Gene Symbol: Ripk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023663.6 | NP_076152.2 | 5, 6 | 888 | ||
| GenBank mRNA | AF302127.3 | - | 5, 6 | 891 | ||
| AK077233.1 | - | 5, 6 | 1072 | |||
| AK145203.1 | - | 5, 6 | 870 | |||
| BC057871.1 | - | 5, 6 | 851 | 5294197 | ||

Ripk4
Genomic Map