siRNA Details
| Gene Symbol: Tmem175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163532.1 | NP_001157004.1 | 3, 4 | 392 | ||
| NM_028223.3 | NP_082499.3 | 3, 4 | 499 | |||
| GenBank mRNA | AI526594.1 | - | 3, 4 | 363 | ||
| AK013879.1 | - | 3, 4 | 322 | |||
| AK038865.1 | - | 3, 4 | 359 | |||
| AK049952.1 | - | 3, 4 | 433 | |||
| AK082765.1 | - | 3, 4 | 355 | |||
| AK170273.1 | - | 3, 4 | 342 | |||
| BB653267.1 | - | 3, 4 | 429 | |||
| BC005542.1 | - | 3, 4 | 307 | 3590497 | ||

Tmem175
Genomic Map