siRNA Details
| Gene Symbol: Lrpprc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028233.2 | NP_082509.2 | 4, 5 | 854 | ||
| GenBank mRNA | AK039751.1 | - | 4, 5 | 630 | ||
| AK138527.1 | - | 4 | 858 | |||
| AK149503.1 | - | 1 | 1053 | |||
| BC059862.1 | - | 4, 5 | 588 | 6417407 | ||
| BC060199.1 | - | 4, 5 | 626 | |||

Lrpprc
Genomic Map