siRNA Details
| Gene Symbol: Lrpprc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028233.2 | NP_082509.2 | 36 | 4212 | ||
| GenBank mRNA | AK013955.1 | - | 17 | 1900 | ||
| BC004681.1 | - | 13 | 1453 | |||
| BC059862.1 | - | 36 | 3946 | 6417407 | ||
| BC060199.1 | - | 36 | 3983 | |||

Lrpprc
Genomic Map