siRNA Details
| Gene Symbol: Ttc30b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028235.1 | NP_082511.1 | 1 | 1997 | ||
| GenBank mRNA | AK011097.1 | - | 1 | 1974 | ||
| AK147134.1 | - | 1 | 1947 | |||
| BC026507.1 | - | 1 | 1189 | |||
| BC151060.1 | - | 1 | 1997 | 9087973 | ||
| BC151184.1 | - | 1 | 1997 | 9088073 | ||
| BC157929.1 | - | 1 | 2066 | 9088102 | ||

Ttc30b
Genomic Map