siRNA Details
| Gene Symbol: Katnbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024254.3 | NP_077216.1 | 8 | 921 | ||
| GenBank mRNA | AK010654.1 | - | 8 | 900 | ||
| AK036861.1 | - | 8 | 921 | |||
| AK150905.1 | - | 1 | 69 | |||
| AK151464.1 | - | 8 | 907 | |||
| AK152570.1 | - | 8 | 886 | |||
| AK168639.1 | - | 8 | 919 | |||
| BC003216.1 | - | 8 | 877 | 3157982 | ||
| BC012402.1 | - | 8 | 877 | 3157982 | ||
| BC080661.1 | - | 8 | 877 | 4460520 | ||

Katnbl1
Genomic Map