siRNA Details
| Gene Symbol: Katnbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024254.3 | NP_077216.1 | 4 | 507 | ||
| GenBank mRNA | AK010654.1 | - | 4 | 486 | ||
| AK036861.1 | - | 4 | 507 | |||
| AK049599.1 | - | 4 | 493 | |||
| AK151464.1 | - | 4 | 493 | |||
| AK152570.1 | - | 4 | 472 | |||
| AK168639.1 | - | 4 | 505 | |||
| BC003216.1 | - | 4 | 463 | 3157982 | ||
| BC012402.1 | - | 4 | 463 | 3157982 | ||
| BC080661.1 | - | 4 | 463 | 4460520 | ||

Katnbl1
Genomic Map