siRNA Details
| Gene Symbol: Ceacam18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028236.1 | NP_082512.1 | 2 | 231 | ||
| GenBank mRNA | AK008389.1 | - | 2 | 263 | ||
| BC042780.1 | - | 2 | 231 | 4224563 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ceacam18| Gene Symbol: Ceacam18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028236.1 | NP_082512.1 | 2 | 231 | ||
| GenBank mRNA | AK008389.1 | - | 2 | 263 | ||
| BC042780.1 | - | 2 | 231 | 4224563 | ||