siRNA Details
Gene Symbol: Spink5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081180.1 | NP_001074649.1 | 28 | 2744 | ||
GenBank mRNA | AJ426449.2 | - | 28 | 2742 | ||
AK010042.1 | - | 1 | 70 | |||
AK028727.1 | - | 9 | 894 | |||
BC118011.1 | - | 28 | 2716 | 40097427 |
