siRNA Details
Gene Symbol: Spink5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081180.1 | NP_001074649.1 | 25 | 2460 | ||
GenBank mRNA | AJ426449.2 | - | 25 | 2458 | ||
AK028727.1 | - | 6 | 610 | |||
BC118011.1 | - | 25 | 2432 | 40097427 |
