siRNA Details
| Gene Symbol: Spink5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081180.1 | NP_001074649.1 | 29 | 2839 | ||
| GenBank mRNA | AJ426449.2 | - | 29 | 2837 | ||
| AK010042.1 | - | 2 | 165 | |||
| AK028727.1 | - | 10 | 989 | |||
| BC118011.1 | - | 29 | 2811 | 40097427 | ||

Spink5
Genomic Map