siRNA Details
| Gene Symbol: Prr5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083810.2 | NP_001077279.1 | 4 | 662 | ||
| NM_001110849.1 | NP_001104319.1 | 5 | 525 | |||
| NM_175181.5 | NP_780390.3 | 4 | 611 | |||
| GenBank mRNA | AK030499.1 | - | 4 | 570 | ||
| AK145981.1 | - | 4 | 613 | |||
| BC048394.1 | - | 4 | 521 | 6391924 | ||
| BC172132.1 | - | 3 | 337 | 9054802 | ||
| BC172133.1 | - | 3 | 337 | |||
| BC172158.1 | - | 3 | 337 | |||
| BY737227.1 | - | 4 | 611 | |||

Prr5l
Genomic Map