siRNA Details
| Gene Symbol: Ccdc71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133744.4 | NP_598505.1 | 2 | 1337 | ||
| GenBank mRNA | AK011229.1 | - | 2 | 1300 | ||
| AK044601.1 | - | 2 | 1303 | |||
| AK079333.1 | - | 2 | 1312 | |||
| AK085544.1 | - | 2 | 1331 | |||
| AK086560.1 | - | 2 | 1303 | |||
| AK136982.1 | - | 2 | 1291 | |||
| AK149878.1 | - | 2 | 1303 | |||
| AK161396.1 | - | 2 | 1413 | |||
| BC018518.1 | - | 2 | 1288 | 2647509 | ||

Ccdc71
Genomic Map