siRNA Details
Gene Symbol: Prcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028243.3 | NP_082519.1 | 9 | 1509 | ||
GenBank mRNA | AK011112.1 | - | 9 | 1488 | ||
AK051677.1 | - | 9 | 1466 | |||
AK054462.1 | - | 9 | 1447 | |||
BC026424.1 | - | 9 | 1305 | |||
BC089575.1 | - | 9 | 1359 |
